Learn More
Faster™ Automatic Regulator with Full Digital Display and Anemometer
Product Code. 16288630
Click to view available options
For Use With (Equipment):
FlowFAST H 09/12
FlowFAST H 15/18
FlowFAST V12, V15, V18, SmartFAST
SmartFAST Laminar Flow Cabinet
Product Code. 16288630
Brand: Faster™ FX0011249800
Factory fitted digital display with anemometer results and ability to control UV lamp.
  • When cabinet is equipped with anemometer with automatic regulation, visualization of air speed and flow alarms will come together
  • Cabinet is supplied with keyboard
Specifications
Includes Automatic Regulator
For Use With (Equipment) SmartFAST Laminar Flow Cabinet