Learn More
BRAND™ Dispenser, tilt measure without bottle, Boro 3.3, NS 29/32
Product Code. 10367582 Shop All Brand Products
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
10367582 5 mL Each
10788764 10 mL Each
10787444 20 mL Each
10205712 25 mL Each
10582073 50 mL Each
10012392 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10367582 Supplier BRAND™ Supplier No. 43055
Only null left
Add to basket
Add to basket
Tilt dispenser is best suited for dispensing suspensions.
  • Dispenser head
  • Calibrated to deliver (TD, Ex)
  • Inscriptions in green enamel
  • Particularly well-suited for dispensing suspensions
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass 3.3
Description Without bottle
Capacity (Metric) 5 mL