missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Autoimmune Regulator/AIRE Polyclonal antibody specifically detects Autoimmune Regulator/AIRE in Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | Autoimmune Regulator/AIRE |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 755 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | APECED protein, APECEDAPSI, Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein, autoimmune regulator, autoimmune regulator (APECED protein)10APS1AIRE1, autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermaldystrophy), PGA1 |
| Host Species | Goat |
| Immunogen | E. coli-derived recombinant mouse Autoimmune Regulator/AIRE, Ser476-Ser552, Accession # Q9Z0E3 |
| Purification Method | Antigen Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?