missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Autoimmune Regulator/AIRE Antibody (609930) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB6184JF635
This item is not returnable.
View return policy
Description
Autoimmune Regulator/AIRE Monoclonal antibody specifically detects Autoimmune Regulator/AIRE in Mouse samples. It is validated for Western Blot,Flow Cytometry (Intracellular Staining),CyTOF
Specifications
| Autoimmune Regulator/AIRE | |
| Monoclonal | |
| Janelia Fluor 635 | |
| APECED protein, APECEDAPSI, Autoimmune polyendocrinopathy candidiasis ectodermal dystrophy protein, autoimmune regulator, autoimmune regulator (APECED protein)10APS1AIRE1, autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermaldystrophy), PGA1 | |
| E. coli-derived recombinant mouse AIRE, Ser476-Ser552 (predicted), Accession # Q9Z0E3 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Western Blot, Flow Cytometry (Intracellular Staining), CyTOF | |
| 609930 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Immune System Diseases, Immunology | |
| 326 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction