Learn More
Cole-Parmer™ PVDF Compression to Male Threaded Adapter Fittings
Product Code. 11739519
Change view
Click to view available options
For Use With:
1/4 tubing O.D. x 1/4 in. NPT(M) Thread
1/4 tubing O.D. x 1/8 in. NPT(M) Thread
3/8 tubing O.D. x 1/4 in. NPT(M) Thread
3/8 tubing O.D. x 3/8 in. NPT(M) Thread
Port Size:
11/32 in. orifice, 1-13/32 x 13/16 in.
5/16 in. orifice, 1-11/32 x 13/16 in.
7/32 in. orifice, 1-3/16 x 5/8 in.
7/32 in. orifice; 1 x 5/8 in.
Thread Size:
1/4 in. NPT(M)
1/8 in. NPT(M)
3/8 in. NPT(M)
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Port Size Thread Size unitSize
11739519 1/4 tubing O.D. x 1/8 in. NPT(M) Thread 7/32 in. orifice; 1 x 5/8 in. 1/8 in. NPT(M) Each
11744749 1/4 tubing O.D. x 1/4 in. NPT(M) Thread 7/32 in. orifice, 1-3/16 x 5/8 in. 1/4 in. NPT(M) Each
11764749 3/8 tubing O.D. x 3/8 in. NPT(M) Thread 11/32 in. orifice, 1-13/32 x 13/16 in. 3/8 in. NPT(M) Each
11754749 3/8 tubing O.D. x 1/4 in. NPT(M) Thread 5/16 in. orifice, 1-11/32 x 13/16 in. 1/4 in. NPT(M) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11739519 Supplier Cole-Parmer™ Supplier No. 0638581
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
These compression fittings are an excellent choice to connect rigid tubing systems, with higher pressure resistance than that provided by barbed fittings.
  • For rigid tubing systems, including tubing made from fluoropolymers, other rigid plastics, and metals
  • Assemble the fitting to tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
  • No leaks, flow restrictions and costly downtime
  • Sterilization by autoclave or ethylene oxide (ETO)
Specifications
Autoclavable Autoclavable
Material PVDF
For Use With 1/4 tubing O.D. x 1/8 in. NPT(M) Thread
Packaging Quantity 1 Each
Port Size 7/32 in. orifice; 1 x 5/8 in.
Sterilization Method Autoclave or Ethylene Oxide
Thread Size 1/8 in. NPT(M)
Product Type Compression Fitting