Learn More
Cole-Parmer™ Parker Hannifin™ PFA Union Connector Compression Fitting
Compression fittings are an excellent choice to connect rigid tubing systems, able to withstand higher pressures than barbed fittings.
Brand:  Cole-Parmer™ 31320-30
Code : A0
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11745788
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Available in straight, "T", and elbow shapes for unmatched versatility
  • Provided with a nut that compresses rigid tubing against the fitting body for high-pressure applications
  • Excellent for use with tubing manufactured from fluoropolymers, other rigid plastics and metals
  • Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal
  • Sterilization through autoclaving or ethylene oxide (ETO)
Specifications
Specifications
Compression Fitting
PFA
1 Each
T-shape
Autoclavable
1/4 in. tubing O.D.
Autoclave or Ethylene Oxide