Learn More
Cole-Parmer™ Parker Hannifin™ PFA Threaded Adapter Compression Fitting
Product Code. 11755099
Change view
Click to view available options
For Use With:
1/2 tubing O.D. x 1/2 in. NPT(M) Thread
1/4 tubing O.D. x 1/4 in. NPT(F) Thread
1/4 tubing O.D. x 1/8 in. NPT(M) Thread
Thread Size:
1/2 in. NPT(M)
1/4 in. NPT(F)
1/8 in. NPT(M)
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With Thread Size unitSize
11755099 1/2 tubing O.D. x 1/2 in. NPT(M) Thread 1/2 in. NPT(M) Each
11740429 1/4 tubing O.D. x 1/4 in. NPT(F) Thread 1/4 in. NPT(F) Each
11795728 1/4 tubing O.D. x 1/8 in. NPT(M) Thread 1/8 in. NPT(M) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11755099 Supplier Cole-Parmer™ Supplier No. 3132013
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings.
  • Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and use the nut to compress down to form a secure seal
  • Superior chemical resistance with durable PFA
  • Available in male and female varieties
  • Sterilization through autoclave or ethylene oxide (ETO)
Specifications
Autoclavable Autoclavable
Material PFA
For Use With 1/2 tubing O.D. x 1/2 in. NPT(M) Thread
Packaging Quantity 1 Each
Sterilization Method Autoclave or Ethylene Oxide
Thread Size 1/2 in. NPT(M)
Product Type Compression Fitting