Learn More
DWK Life Sciences DURAN™ Tube with Screw Thread, with DIN thread, and standard ground cone
The threaded tube with thread and standard ground joint allows the bonding of various glass components.
Brand: DWK Life Sciences 248408203
Code : Z4
Additional Details : Weight : 0.17000kg
Description
This threaded tube has a screw thread and a standard ground joint. Components of an apparatus can be connected via both ends and thus connected to each other.
- Tight closure due to standard ground joint
- With DIN thread
Specifications
10/Pk. | |
Tube with screw threads | |
Non-DIN Size | |
GL 14, with Cone NS 24/29 |