missing translation for 'onlineSavingsMsg'
Learn More

ratiolab™ Autoclavable Contaminated Labware Bags

Product Code. 16621612
Change view
Click to view available options
Dimensions (L x W):
290 x 210 mm
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
16621612 290 x 210 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16621612 Supplier ratiolab™ Supplier No. 7002110
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Distinctive red autoclavable bags for easy identification, for the disposal of contaminated labware.

  • With BIOHAZARD-Warning Instruction and sterilization indicator patch
  • Safety imprint in German, English, French and Spanish languages
  • Rectangle white indicator field turns into dark when autoclaved
  • For autoclaving, the bag must not be completely closed
  • Autoclavable at 121°C for 20 min.

Specifications

Autoclavable Autoclavable
Color Red
Disposable Single-use
For Use With (Application) Waste Disposal
Material Polyethylene
Quantity 100 Plates/Box, 10 Boxes/Pk.
Dimensions (L x W) 290 x 210 mm
Product Type Biohazard Disposal Bag
Capacity (English) 0.4 gal.
Capacity (Metric) 1.5 L
Thickness (Metric) 20 μm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.