Learn More
Abeba™ Autoclavable Clogs 9620
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
Unit Size:
1 pair
Description
- CE EN ISO 20347:2012 OB A, E SRC
- Autoclavable, sterilisable special material
- Green
- Antistatic
- Heel strap foldable, adjustable
- DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications
Specifications
| Certifications/Compliance | CE EN ISO 20347:2012 OB A, E SRC |
| Color | Green |
| Size | 41 |
| Style | 9620 |