missing translation for 'onlineSavingsMsg'
Learn More

Abeba™ Autoclavable Clogs 39620

Antistatic and autoclavable at 135°C for 5 minutes. High slip resistance. Washing at 60°C.

Brand:  Abeba™ 39620/43

Additional Details : Weight : 0.55000kg

 View more versions of this product

Product Code. 15691258

  • £76.59 / 1 pair
Estimated Shipment: 11-03-2024
to see stock.
Add to basket

Description

Description

  • CE EN ISO 20347:2012 OB A, E SRC
  • Autoclavable, sterilisable special material
  • Green
  • Slip resistance outsole sole
  • ESD dissipative according to DIN EN 61340
  • Heel strap foldable, adjustable
  • DOUBLE SIZES, please use even size when placing your order (36, 38, 40, 42, 44, 46)
Specifications

Specifications

CE EN ISO 20347:2012 OB A, E SRC
43
39620
Green
Slip-resistant
Recommendations

Recommendations

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers