missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Autocamtide-2-related inhibitory peptide, myristoylated

Description
Autocamtide-2-related inhibitory peptide, myristoylated is a CaM kinase II inhibitor; enhanced cell permeable derivative of Autocamtide-2-related inhibitory peptide (Cat No. 1688). Pretreatment blocks reinstatement of morphine-seeking behavior in vivo.
Specifications
Specifications
| Purity | 0.95 |
| CAS | 201422-04-0 |
| Content And Storage | Store at -20°C |
| Description | CaM Kinase Inhibitors |
| Molecular Weight (g/mol) | Observed M.W.: 1708.12 |
| Quantity | 1 mg |
| Structural Form | C78H142N22O20 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?