missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals AutoBlot System 36

Product Code. 30694938 Shop All MP Biomedicals Products
Change view
Click to view available options
Capacity:
36 Strips
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
30694938 36 Strips Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30694938 Supplier MP Biomedicals Supplier No. EMC019

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

AutoBlot System is an intelligent Western Blot processor that enables automated washing and incubation of up to 36 assay strips per run at your pre-selected temperature.

  • AutoBlot System 36 is an automated Western Blot instrument that delivers automatic operation for Western blotting membrane strip experiment.
  • Add sample according to experiment requirements, the instrument then automatically dispenses liquid, aspirates liquid, incubates, rinses, and adds substrate.
  • Liquid aspiration volume, distribution volume, time, temperature and shaking speed of incubation plate, can be set or edited by the user.
  • AutoBlot System 36 is composed of an instrument, dispensing and washing bottles, waste bottles, disposable incubation plate of 36 strips and control software.
  • This system helps to satisfy your Immunoblot needs by minimizing human error, increasing productivity and delivering accurate and consistent results.
  • CE-marked.
  • For professional use only.
  • Reduce work load.
  • Run an entire batch of samples in a single operation.

Specifications

Capacity 36 Strips
Detection Method Colorimetric
For Use With (Application) Western Blot
For Use With (Equipment) AutoScan Pro
Gel Compatibility N/A
Gel Size N/A
Membrane Compatibility Nitrocellulose
Mode of Transfer N/A
Running Time 3 to 4 hr.
Throughput 36 Strips/Run
Type Blot Processor
Gel Dimensions No Gel Required
Gel Type No Gel Required
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.