Learn More
Thermo Scientific™ Auto-refill for VersaCool™ Refrigerated Circulating Bath
Automatically maintain and ensure the optimal level of fluid in your VersaCool with the Autorefill feature.
Brand:  Thermo Scientific™ 230000005
Additional Details : Weight : 2.00000kg
Product Code. 15419539
  • £850.16 / Each
Estimated Shipment: 12-07-2024
to see stock.
Add to basket
Description
Description
Automatically maintain and ensure the optimal level of fluid in your VersaCool with the Autorefill feature. Provides peace of mind with user-defined start and stop levels, as well as a time-out feature. The needle valve allows control of the fill rate from as fast a refill as possible- to a slow refill that minimizes any effect on temperature stability or any rate in between
Specifications
Specifications
Auto-refill for VersaCoolandtrade Refrigerated Circulating Bath
Thermo Scientific VersaCool Refrigerated Circulating Bath
Auto-Refill Device