missing translation for 'onlineSavingsMsg'
Learn More

AUT1, MedChemExpress

Product Code. 16471137
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471137 50 mg 50mg
16457637 5 mg 5mg
16445666 100 mg 100mg
16434888 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16471137 Supplier Medchem Express Supplier No. HY117639
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AUT1 is a Kv3 potassium channel modulator, with pEC50s of 5.33 and 5.31 for human recombinant Kv3.1b and Kv3.2a, respectively, exhibits 10-fold lower potency at human recombinant Kv3.3 channel (pEC50, 4.5).

AUT1 is a Kv3 potassium channel modulator, with pEC50s of 5.33 and 5.31 for human recombinant Kv3.1b and Kv3.2a, respectively, exhibits 10-fold lower potency at human recombinant Kv3.3 channel (pEC50, 4.5).

Specifications

Chemical Name or Material AUT1
CAS 1311136-84-1
Color White
Purity Grade Notes Research
Molecular Formula C18H19N3O4
Quantity 50 mg
Solubility Information DMSO : ≥ 250 mg/mL (732.36 mM)
SMILES O=C1N(C2=CC=C(OC3=CC=C(C)C(OC)=C3)N=C2)C([C@@H](CC)N1)=O
Molecular Weight (g/mol) 341.36
Formula Weight 341.36
Percent Purity 99.84%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.