missing translation for 'onlineSavingsMsg'
Learn More

Aurora C Antibody (OTI2B7), PE, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-71303PE

Product Code. 30028256

  • £561.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Aurora C Monoclonal antibody specifically detects Aurora C in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aurora C
Monoclonal
PE
PBS
Mouse
Immunogen affinity purified
RUO
Primary
Human
Purified
Western Blot, Immunohistochemistry
OTI2B7
Western Blot, Immunohistochemistry
AIE2AIK3, ARK3Serine/threonine-protein kinase aurora-C, AurC, aurora kinase CAurora/IPL1/Eg2 protein 2, Aurora/IPL1-related kinase 3, aurora-C, Aurora-related kinase 3, EC 2.7.11, EC 2.7.11.1, serine/threonine kinase 13 (aurora/IPL1-like), serine/threonine-protein kinase 13, STK13ARK-3
Full-length protein expressed in 293T cell transfected with human AURKC expression vector
0.1 mL
Cell Cycle and Replication
6795
Store at 4C in the dark.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.