missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aurora B Antibody (AURKB/3121R), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-08280AF700
This item is not returnable.
View return policy
Description
Aurora B Monoclonal antibody specifically detects Aurora B in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Aurora B | |
| Monoclonal | |
| Alexa Fluor 700 | |
| AIK2EC 2.7.11.1, AIM1Aik2, AIM-1STK-1, ARK2ARK-2, AurB, aurkb-sv2, Aurora- and IPL1-like midbody-associated protein 1, aurora kinase BSerine/threonine-protein kinase aurora-B, aurora kinase B-Sv1, aurora kinase B-Sv2, Aurora/IPL1-related kinase 2, aurora-1, aurora-B, Aurora-related kinase 2, EC 2.7.11, IPL1, serine/threonine kinase 12, serine/threonine-protein kinase 12, STK12aurkb-sv1, STK5 | |
| Recombinant fragment (around aa 89-251) of human Aurora B protein (exact sequence is proprietary) (Uniprot: Q96GD4) | |
| 100 μg | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Stem Cell Markers | |
| 9212 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| AURKB/3121R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction