missing translation for 'onlineSavingsMsg'
Learn More

Aurora B Antibody (AURKB/1521), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18421673 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421673 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421673 Supplier Novus Biologicals Supplier No. NBP254478AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Aurora B Monoclonal antibody specifically detects Aurora B in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aurora B
Applications Western Blot
Classification Monoclonal
Clone AURKB/1521
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias AIK2EC 2.7.11.1, AIM1Aik2, AIM-1STK-1, ARK2ARK-2, AurB, aurkb-sv2, Aurora- and IPL1-like midbody-associated protein 1, aurora kinase BSerine/threonine-protein kinase aurora-B, aurora kinase B-Sv1, aurora kinase B-Sv2, Aurora/IPL1-related kinase 2, aurora-1, aurora-B, Aurora-related kinase 2, EC 2.7.11, IPL1, serine/threonine kinase 12, serine/threonine-protein kinase 12, STK12aurkb-sv1, STK5
Host Species Mouse
Immunogen Recombinant fragment (around aa 89-251) of human Aurora B protein (exact sequence is proprietary) (Uniprot: Q96GD4)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 9212
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.