missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aurora B Antibody (3F11), Novus Biologicals™
Mouse Monoclonal Antibody
£146.00 - £395.00
Specifications
| Antigen | Aurora B |
|---|---|
| Clone | 3F11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000, Proximity Ligation Assay |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Proximity Ligation Assay |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18661999
|
Novus Biologicals
NBP2-50039SS |
0.025 mL |
£146.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648798
|
Novus Biologicals
NBP2-50039 |
0.1 mL |
£395.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aurora B Monoclonal antibody specifically detects Aurora B in Human, Mouse, Rat, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Proximity Ligation AssaySpecifications
| Aurora B | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Proximity Ligation Assay | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, DNA Repair, Mitotic Regulators, Stem Cell Markers | |
| 50% PBS, 50% glycerol | |
| AIK2EC 2.7.11.1, AIM1Aik2, AIM-1STK-1, ARK2ARK-2, AurB, aurkb-sv2, Aurora- and IPL1-like midbody-associated protein 1, aurora kinase BSerine/threonine-protein kinase aurora-B, aurora kinase B-Sv1, aurora kinase B-Sv2, Aurora/IPL1-related kinase 2, aurora-1, aurora-B, Aurora-related kinase 2, EC 2.7.11, IPL1, serine/threonine kinase 12, serine/threonine-protein kinase 12, STK12aurkb-sv1, STK5 | |
| Full length recombinant human aurora B protein expressed in and purified from E. coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 3F11 | |
| Western Blot 1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000, Proximity Ligation Assay | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Bovine, Equine | |
| Q96GD4, Q96GD4 | |
| 9212 | |
| IgG2a | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title