missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aurora A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10297-1-AP-150UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a cell cycle-regulated kinase that appears to be involved in microtubule formation and/or stabilization at the spindle pole during chromosome segregation. The encoded protein is found at the centrosome in interphase cells and at the spindle poles in mitosis. This gene may play a role in tumor development and progression. A processed pseudogene of this gene has been found on chromosome 1, and an unprocessed pseudogene has been found on chromosome 10. Multiple transcript variants encoding the same protein have been found for this gene.Specifications
| Aurora A | |
| Polyclonal | |
| Unconjugated | |
| AURKA | |
| AIK, ARK 1, ARK1, AURA, AURKA, Aurora A, aurora kinase A, Aurora related kinase 1, Aurora/IPL1 related kinase 1, AURORA2, Breast tumor amplified kinase, BTAK, hARK1, STK15, STK6, STK7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 20878, 261730, 6790 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O14965, P59241, P97477 | |
| AURKA | |
| Aurora A Fusion Protein Ag0362 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction