Learn More
Eurofins DiscoverX Aurora A Protein, Active
Description
Protein serine/threonine kinase Aurora-A (STK6, STK15, BTAK, AIK, ARK1, AURA, Aurora-related kinase 1, Breast-tumor-amplified kinase) is one of a family of serine-threonine kinases that have been strongly linked to chromosome segregation and centrosome functions. There are three isoforms of Aurora kinase: A, B, and C. Molecular mechanism of Aurora-A regulation of G2-M transition is phosphorylation of histone H3. Inhibition of Aurora A may be useful in the treatment of many types of human cancer, including leukemia, colon and breast cancer.
GenBank NM_003600. The recombinant protein also contains the conflict I31F with respect toGenBank NM_003600. This substitution is reported inGenBank NM_003158 and BC027464. The residue coordinates in the native sequence are given.
Specifications
Specifications
| Formulation | mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
| Molecular Weight (g/mol) | 46.9 kDa |
| Quantity | 250 μg |
| Species | Human |
| Recombinant | Recombinant |
| Protein Tag | N-terminal 6His |
| Expression System | Recombinant enzyme expressed in Sf21 insect cells |
| Protein Form | Full-length |
| Purity or Quality Grade | ≥50% |
| Protein | Aurora A, active |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.