missing translation for 'onlineSavingsMsg'
Learn More

Aurora A/B Antibody (3H1), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18770260 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18770260 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18770260 Supplier Novus Biologicals Supplier No. NBP250043AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Aurora A/B Monoclonal antibody specifically detects Aurora A/B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aurora A/B
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 3H1
Conjugate Alexa Fluor 405
Formulation 50 mM Sodium Borate
Gene Alias AIK, AIK2, AIM1, AIM-1, AIRK1, AIRK2, ARK1, ARK2, AURA, AurB, AURKA, AURKB, aurkb-sv1, aurkb-sv2, AYK1, BTAK, IAK1, IPL1, PPP1R47, PPP1R48, STK1, STK12, STK15, STK5, STK6, STK7
Host Species Mouse
Immunogen Recombinant full length human Aurora A expressed in and purified from E. coli. [UniProt# O14965]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.