missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aurora A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 6 publications
Brand: Novus Biologicals NBP1-51843
This item is not returnable.
View return policy
Description
Aurora A Polyclonal specifically detects Aurora A in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Aurora A | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| AIKAurA, ARK-1, ARK1EC 2.7.11.1, AURA, aurora kinase AhARK1, aurora/IPL1-like kinase, Aurora/IPL1-related kinase 1, Aurora-related kinase 1, Breast tumor-amplified kinase, breast-tumor-amplified kinase, BTAKAURORA2, EC 2.7.11, IPL1-related kinase, serine/threonine kinase 6, serine/threonine protein kinase 15, Serine/threonine-protein kinase 15, serine/threonine-protein kinase 6, Serine/threonine-protein kinase aurora-A, STK15serine/threonine kinase 15, STK6MGC34538, STK7 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/ml | |
| Western Blot 1 - 2 ug/ml, Simple Western 1:200, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 2 ug/ml, Immunohistochemistry-Paraffin 1:200 | |
| O14965, P97477 | |
| AURKA | |
| A recombinant protein made to an N-terminal region of the human Aurora A protein (within residues 50-200). [Swiss-Prot O14965] | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, MAP Kinase Signaling, Mitotic Regulators, Protein Kinase, Signal Transduction, Stem Cell Markers, Tumor Suppressors | |
| 6790 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction