missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aurora A Antibody (1F8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-22118
This item is not returnable.
View return policy
Description
Aurora A Monoclonal specifically detects Aurora A in Human, Rat, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Aurora A | |
| Monoclonal | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| AIKAurA, ARK-1, ARK1EC 2.7.11.1, AURA, aurora kinase AhARK1, aurora/IPL1-like kinase, Aurora/IPL1-related kinase 1, Aurora-related kinase 1, Breast tumor-amplified kinase, breast-tumor-amplified kinase, BTAKAURORA2, EC 2.7.11, IPL1-related kinase, serine/threonine kinase 6, serine/threonine protein kinase 15, Serine/threonine-protein kinase 15, serine/threonine-protein kinase 6, Serine/threonine-protein kinase aurora-A, STK15serine/threonine kinase 15, STK6MGC34538, STK7 | |
| Mouse | |
| 48 kDa | |
| 0.1 mL | |
| Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, MAP Kinase Signaling, Mitotic Regulators, Protein Kinase, Signal Transduction, Stem Cell Markers, Tumor Suppressors | |
| 6790 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1F8 | |
| Unconjugated | |
| O14965 | |
| AURKA | |
| Purified recombinant fragment of human Aurora A expressed in E. coli. [Uniprot: O14965] | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Rat, Primate | |
| Precipitation |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction