missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human AURKB (Aurora B), His Tag Recombinant Protein

Product Code. 10680571
Click to view available options
Quantity:
10 μg
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10680571

Brand: Invitrogen™ PV6130

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length.

The Aurora kinases associate with microtubules during chromosome movement and segregation. AURKB (Aurora B) localizes to microtubules near kinetochores, specifically to the specialized microtubules called K-fibers. This complex functions in chromosome alignment and separation, histone modification, and cytokinesis. CPC also includes three non-enzymatic subunits known as the inner centromere protein (INCENP), survivin, and borealin, which determine the activity, localization, stability, and possibly even the substrate specificity of AurB. Two CPCs exist during mitosis: one containing all four members and another consisting of INCENP and AurB. Quaternary CPC functions during chromosome alignment and cytokinesis, whereas the INCENP-AurB complex may be responsible for modifying histone H3. AurB is overexpressed in a number of tumours, including primary breast and colon tumour samples. Its expression in tumours seems to be linked with Aurora-A kinase, suggesting a feedback mechanism between the two proteins.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96GD4
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris with 50% glycerol, 0.02% Triton X-100, 150mM NaCl, 0.5mM EDTA, 2mM DTT and no preservative; pH 7.5
Gene ID (Entrez) 9212
Molecular Weight (g/mol) 43.4 kDa
Name Human AURKB (Aurora B), His Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Aik2; AIM1; AIM-1; AIRK2; AL022959; ARK2; ARK-2; AurB; AURKB; aurkb-sv1; aurkb-sv2; aurora 1; aurora- and IPL1-like midbody-associated protein 1; aurora B; aurora kinase B; aurora kinase B-Sv1; aurora kinase B-Sv2; aurora/IPL1-related kinase 2; aurora-1; aurora-B; aurora-related kinase 2; IPL1; PPP1R48; protein phosphatase 1, regulatory subunit 48; serine/threonine kinase; serine/threonine kinase 12; serine/threonine kinase 5; Serine/threonine-protein kinase 12; Serine/threonine-protein kinase 5; serine/threonine-protein kinase aurora-B; STK1; STK-1; Stk12; STK12; AIK2; AIM1; ARK2; IPL1; AIM-1; STK5
Common Name Aurora B
Gene Symbol AURKB
KinaseFamily Aurora Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Kinase Group Ser⁄Thr Kinases: Other
Shipping Condition Dry Ice
System Compatibility LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate
Target Entry AURKB
Target Gene aurora kinase B
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Signal Transduction
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.