missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AURKAIP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550356
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total AURKAIP1 protein.
AURKA is a cell cycle-regulated kinase that appears to be involved in microtubule formation and/or stabilization at the spindle pole during chromosome segregation. The encoded protein is found at the centrosome in interphase cells and at the spindle poles in mitosis. This gene may play a role in tumor development and progression.
Specifications
| AURKAIP1 | |
| Polyclonal | |
| Unconjugated | |
| AURKAIP1 | |
| 0610033H09Rik; 28S ribosomal protein S38, mitochondrial; Aip; AKIP; AURKA; AURKA-interacting protein; AURKAIP1; aurora kinase A interacting protein 1; aurora kinase A-interacting protein; aurora-A kinase interacting protein; Mitochondrial small ribosomal subunit protein mS38; Mrps38; MRP-S38 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54998 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 2.6 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9NWT8 | |
| AURKAIP1 | |
| Full length fusion protein of Human AURKAIP1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction