Learn More
AURKA Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28371-1-AP-20UL
This item is not returnable.
View return policy
Description
The AURKA antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human AURKA. This antibody recognizes human antigen. The AURKA antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| AURKA | |
| 1400 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, ELISA | |
| AIK, ARK 1, ARK1, AURA, AURKA, aurora kinase A, Aurora related kinase 1, Aurora/IPL1 related kinase 1, AURORA2, Breast tumor amplified kinase, BTAK, hARK1, STK15, STK6, STK7 | |
| 20 μL | |
| 6790 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |