missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human AURKA (Aurora A), His Tag Recombinant Protein

Product Code. 11573467
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 11573467

Brand: Invitrogen™ PV3612

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length Assay Entry: Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic System Compatibility: LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate.

AURKA (Aurora A) is a serine/threonine kinase that regulates mitosis by association in vivo with centrosomes. Overexpression of AURKA is observed in many types of cancer. Aurora A plays a role in cell cycle regulation during anaphase and/or telophase, in relation to the function of the centrosome/spindle pole region during chromosome segregation. Aurora A plays a key role during tumor development and progression and is overexpressed in many human cancers including breast, ovarian and colorectal. Aurora A is viewed as a potential target for anticancer drug treatment. Tissue specificity: Highly expressed in testis and weakly in skeletal muscle, thymus and spleen.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O14965
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50mM tris HCl with 4mM DTT, 150mM NaCl, 0.5mM EDTA, 0.05% Triton X-100, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 6790
Molecular Weight (g/mol) 49.9 kDa
Name Human AURKA (Aurora A), His Tag
pH Range 7.5
Purification Method Purified
Quantity 10 μg
Storage Requirements -80°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias Aik; Airk; AIRK1; Ark1; ARK-1; AU019385; AURA; AURAK; AURKA; Aurora 2; aurora A; Aurora A Kinase; aurora family kinase 1; Aurora kinase A; aurora/IP; aurora/IPL1-like kinase; aurora/IPL1-related kinase 1; AURORA2; Aurora-A; aurora-related kinase 1; AW539821; Ayk1; Breast tumor-amplified kinase; BTAK; EC 2.7.11.1; hARK1; IAK; IAK1; Ipl1- and aurora-related kinase 1; IPL1-related kinase; MGC34538; OTTHUMP00000031341; OTTHUMP00000166072; PPP1R47; protein phosphatase 1, regulatory subunit 47; ratAurA; RP5-1167H4.6; sereine threonine kinase; serine/threonine kinase 6; serine/threonine protein kinase 15; serine/threonine protein kinase 6; serine/threonine-protein kinase 15; Serine/threonine-protein kinase 6; serine/threonine-protein kinase 6; aurora kinase A; serine/threonine-protein kinase aurora-A; Serine/threonine-protein kinase Ayk1; STK15; STK6; STK7; Unknown (protein for MGC:134472)
Common Name Aurora A
Gene Symbol AURKA
KinaseFamily Aurora Kinase Family
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Assay Entry Biochemical phospho-specific activity (no antibody), Biochemical terbium phospho-specific activity, Biochemical binding/kinetic
Kinase Group Ser⁄Thr Kinases: Other
Shipping Condition Dry Ice
System Compatibility LanthaScreen™ Antibody, LanthaScreen™ Substrate, Z'-LYTE™, Omnia™ Bulk Substrate
Target Entry AURKA
Target Gene aurora kinase A
Expression System Insect cells
Protein Family Kinases & Inhibitors
Protein Form Recombinant, Full Length
Form Liquid
Protein Subtype Serine/Threonine Kinases
Research Category Oncology, Signal Transduction
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.