missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Auramine O, pure, biological stain

Product Code. 10460311 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10460311 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10460311 Supplier Thermo Scientific Chemicals Supplier No. 227520500
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2465-27-2 | C17H22ClN3 | 303.834 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point >250.0°C
CAS 2465-27-2
Dye Content 60% min.
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C17H22ClN3
MDL Number MFCD00012484
Synonym Basic Yellow 2, 4, 4'-(Imidocarbonyl)bis(N, N-dimethylaniline) monohydrochloride, C.I. 41000
Color Ochre/Yellow to Yellow
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in dimethylformamide, acetone and ethanol
Quantity 50 g
InChI Key KSCQDDRPFHTIRL-UHFFFAOYSA-N
Physical Form Powder
SMILES CN(C)C1=CC=C(C=C1)C(=N)C2=CC=C(C=C2)N(C)C.Cl
IUPAC Name 4-[4-(dimethylamino)benzenecarboximidoyl]-N,N-dimethylaniline;hydrochloride
Molecular Weight (g/mol) 303.834
PubChem CID 17170
ChEBI CHEBI:51876
Formula Weight 303.83
Grade Pure
Chemical Name or Material Auramine O
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Auramine O
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 2
  • Acute toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H351-Suspected of causing cancer.
  • H311-Toxic in contact with skin.
Precautionary Statement
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.