missing translation for 'onlineSavingsMsg'
Learn More

Auramine O Hydrochloride (~75%), TRC

Product Code. 18021630
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18021630 1 g 1g
18041020 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18021630 Supplier Toronto Research Chemicals Supplier No. TRCA7945451G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2465-27-2
Molecular Formula C17 H21 N3 . Cl H
Molecular Weight (g/mol) 303.83
Synonym 4,4'-Carbonimidoylbis[N,N-dimethylbenzenamine monohydrochloride, C.I. Basic Yellow 2, C.I. Basic Yellow 2 monohydrochloride, ADC Auramine O, Aizen Auramine, Aizen Auramine OH, Aizen Auramine OW 100, Aizen Auramine conc.SFA, Arazine Yellow, Auramin, Auramine, Auramine 0-100, Auramine A1, Auramine Extra, Auramine Extra 0-100, Auramine Extra 0-125, Auramine Extra Conc. A, Auramine FA, Auramine FWA, Auramine II, Auramine Lake Yellow O, Auramine N, Auramine O, Auramine O Extra Conc. A Export, Auramine ON, Auramine OO, Auramine OOO, Auramine OS, Auramine Pure, Auramine SP, Auramine Yellow, Auramine chloride, Auramine hydrochloride, Basic Flavine Yellow O, Basic Light Yellow, Basic Light Yellow O, Basic Yellow 2, Basic yellow O, Basonyl Yellow 120, C.I. 41000, Calcozine Yellow OX, Flexo Yellow 110, Mitsui Auramine O
IUPAC Name 4-[4-(dimethylamino)benzenecarboximidoyl]-N,N-dimethylaniline;hydrochloride
SMILES Cl.CN(C)c1ccc(cc1)C(=N)c2ccc(cc2)N(C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C17H21N3.ClH/c1-19(2)15-9-5-13(6-10-15)17(18)14-7-11-16(12-8-14)20(3)4;/h5-12,18H,1-4H3;1H
Formula Weight 303.1502
Chemical Name or Material Solvent Yellow 34
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.