missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Auramine O, 80% dye content

Product Code. 15423129
Change view
Click to view available options
Quantity:
25 g
50 g
Unit Size:
25g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15423129 25 g 25g
15433129 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15423129 Supplier Thermo Scientific Chemicals Supplier No. J60343.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Fluorescent stain for acid-resistant bacteria in sputum | CAS: 2465-27-2 | C17H22ClN3 | 303.834 g/mol

Fluorescent stain for acid-resistant bacteria in sputum

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2465-27-2
Molecular Formula C17H22ClN3
Molecular Weight (g/mol) 303.834
MDL Number MFCD00012484
InChI Key KSCQDDRPFHTIRL-UHFFFAOYSA-N
Synonym C.I. 41000
PubChem CID 17170
ChEBI CHEBI:51876
IUPAC Name 4-[4-(dimethylamino)benzenecarboximidoyl]-N,N-dimethylaniline;hydrochloride
SMILES CN(C)C1=CC=C(C=C1)C(=N)C2=CC=C(C=C2)N(C)C.Cl

Specifications

Melting Point 250°C (decomposition)
UN Number UN2811
Beilstein 4030061
Solubility Information Soluble in water or ethanol
Formula Weight 303.84
Color Yellow
Quantity 25 g
Physical Form Powder
Chemical Name or Material Auramine O, 80% dye content

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.