Learn More
3M™ Aura™ Particulate Respirator, 9300+ Gen3 Series
Product Code. 16260262
Click to view available options
Filter Type:
FFP2
FFP3
Assigned Protection Factor (APF):
10
20
With Exhalation Valve:
No
Yes
Quantity:
1/Pk.
10/Pk.
20/Pk.
330/Pk.
5/Pk.
missing translation for 'unitSize'
Pack of 10
Pack of 20
Pack of 330
Pack of 40
Pack of 5
Product Code. 16260262
missing translation for 'mfr': 3M™ 7100138120
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Aura™ Particulate Respirator 9300+Gen3 Series is the next generation of 3M Aura Respirator: FFP protection combined with the original 3-panel, flat-fold design.
  • Original 3-panel, flat-fold design fits a wide range of face shapes and sizes
  • 3M™ Cool Flow™ Comfort Valve allows a greater percentage of warm, exhaled air to escape, helping to increase your comfort compared to 3M™ Aura™ 9300+ equivalent model
  • 3M™ Advanced Electret (filter) Media helps you breathe easier
  • Innovative tabs on the upper and lower panels simplify positioning and adjustment
  • Braided headbands are colour-coded (blue for FFP2, red for FFP3) for quick and easy identification
  • Embossed upper panel reduces eyewear fogging
  • Easy to store away with flat-fold design and individual wrapping
TRUSTED_SUSTAINABILITY
Specifications
Assigned Protection Factor (APF) 10
Certifications/Compliance CE Approved
Certifications/Compliance (Europe) EN 149:2001 + A1:2009, CE Approved
Color White
Design Type Flat-fold
Disposable Single-use
Filter Type FFP2
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size Standard
With Exhalation Valve Yes
Latex Free Yes
Material Headstrap Polyisoprene
Model 9322+Gen3
Nuisance Odor Relief Up to 12 x TLV
Quantity 5/Pk.
Sterility Non-sterile
Style Flat-fold
Weight (Metric) 117 g
FDA Cleared Surgical Mask No