Learn More
Invitrogen™ AUP1 Polyclonal Antibody
Sheep Polyclonal Antibody
Brand: Invitrogen™ PA118257
Description
Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.
AUP1 contains a domain with homology to the ancient conserved region of the archain 1 gene and a domain they may be involved in binding ubiquitin-conjugating enzymes. The unprocessed precursor is of 476 amino acids in length and has an estimated molecular weight of 53 kDa.
Specifications
| AUP1 | |
| Polyclonal | |
| Unconjugated | |
| AUP1 | |
| AA589454; ancient ubiquitous protein 1; ancient ubiquitous protein 1 isoform 2; ancient ubiquitous protein 1; LOW QUALITY PROTEIN: ancient ubiquitous protein 1; Aup1; AUP1 lipid droplet regulating VLDL assembly factor; AUP1, lipid droplet regulating VLDL assembly factor; HUBCEP52; LOW QUALITY PROTEIN: ancient ubiquitous protein 1; RPL40 | |
| Sheep | |
| 100 μL | |
| Primary | |
| Human, Rat, Bovine, Sheep | |
| Antibody | |
| IgG |
| Immunohistochemistry (Frozen), Western Blot | |
| Conc. Not Determined | |
| Whole serum with no preservative | |
| A1L134, Q9Y679 | |
| AUP1 | |
| Synthetic peptide corresponding to residues H(48) V F L V S C A L P D S V(60) of human ubiquitin. | |
| RUO | |
| 101119707, 509130, 550, 680423 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.