missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AUF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | AUF1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232206
|
Novus Biologicals
NBP3-35586-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30230672
|
Novus Biologicals
NBP3-35586-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
AUF1 Polyclonal antibody specifically detects AUF1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ImmunofluorescenceSpecifications
| AUF1 | |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 3184 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AUF1A, heterogeneous nuclear ribonucleoprotein D (AU-rich element RNA binding protein1, 37kDa), heterogeneous nuclear ribonucleoprotein D (AU-rich element RNA-binding protein1, 37kD), heterogeneous nuclear ribonucleoprotein D0, hnRNPD0, type A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-306 of human AUF1 (NP_002129.2).,, Sequence:, MSEEQFGGDGAAAAATAAVGGSAGEQEGAMVAATQGAAAAAGSGAGTGGGTASGGTEGGSAESEGAKIDASKNEEDEGHSNSSPRHSEAATAQREEWKMFIGGLSWDTTKKDLKDYFSKFGEVVDCTLKLDPITGRSRGFGFVLFKESESVDKVMDQKEHKLNGKVIDPKRAKAMKTKEPVKKIFVGGLSPDTPEEKIREYFGGFGEVESIELPMDNKTNKRRGFCFITFKEEEPVKKIMEKKYHNVGLSKCEIKVAMSKEQYQQQQQWGSRGGFAGRARGRGGDQQSGYGKVSRRGGHQNSYKPY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title