missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aubergine/Sting Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-21661
This item is not returnable.
View return policy
Description
Aubergine/Sting Polyclonal specifically detects Aubergine/Sting in Drosophila samples. It is validated for Western Blot, ELISA.
Specifications
| Aubergine/Sting | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| Aub, aubergine, CG6137, Dmel\CG6137, ms(2)32D, sti, sting | |
| Rabbit | |
| 98.56 kDa | |
| 0.1 mg | |
| Primary | |
| Reactive in Drosophila melanogaster. A BLAST analysis was used to suggest limited cross reactivity with Aubergine protein from other sources. | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 1 mg/ml | |
| Western Blot 1:1000-1:10000, ELISA 1:20000 | |
| O76922 | |
| aub | |
| This affinity purified Aubergine/Sting Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of Drosophila melanogaster Aubergine/Sting protein. (Uniprot: O76922) | |
| Affinity Purified | |
| RUO | |
| 34524 | |
| Drosophila | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction