missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AU1235, MedChemExpress
Description
AU1235, an adamantyl urea, is a potent MmpL3 inhibitor. The Mycobacterium tuberculosis protein MmpL3 performs an essential role in cell wall synthesis, since it effects the transport of trehalose monomycolates across the inner membrane.
Specifications
Specifications
| Chemical Name or Material | AU1235 |
| CAS | 1338780-86-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C17H19F3N2O |
| Quantity | 5 mg |
| Solubility Information | DMSO : 25 mg/mL (77.08 mM; Need ultrasonic) |
| SMILES | O=C(NC(C=CC(F)=C1F)=C1F)NC2(C[C@H](C3)C4)C[C@H]4C[C@H]3C2 |
| Molecular Weight (g/mol) | 324.34 |
| Formula Weight | 324.34 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?