missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATXN7L3B Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566027
This item is not returnable.
View return policy
Description
Immunogen sequence: VKCGYFYLEFA ETGSVKDFGI QPVEDKGACR LPLCSLPGEP GNGPDQQLQR SPPEFQ Highest antigen sequence identity to the following orthologs - mouse 89%, rat 93%.
By binding to ENY2, interferes with the nuclear functions of the deubiquitinase (DUB) module of the SAGA complex which consists of ENY2, ATXN7, ATXN7L3 and the histone deubiquitinating component USP22. Affects USP22 DUB activity toward histones indirectly by changing the subcellular distribution of ENY2 and altering ENY2 availability for ATXN7L3 interaction. Regulates H2B monoubiquitination (H2Bub1) levels through cytoplasmic sequestration of ENY2 resulting in loss of nuclear ENY2-ATXN7L3 association which destabilizes ATXN7L3. Affects protein expression levels of ENY2 and ATXN7L3.
Specifications
| ATXN7L3B | |
| Polyclonal | |
| Unconjugated | |
| ATXN7L3B | |
| 4921506J03Rik; 6230409E21Rik; AI315132; ataxin 7 like 3B; ataxin 7-like 3B; Ataxin-7-like protein 3B; ATXN7L3B; ENSMUSG00000074747; lnc-SCA7; putative ataxin-7-like protein 3B; putative ataxin-7-like protein 3-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 552889 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96GX2 | |
| ATXN7L3B | |
| Recombinant Human ATXN7L3B. Recombinant protein control fragment (Product #RP-106717). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction