missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATXN7L2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | ATXN7L2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ATXN7L2 Polyclonal specifically detects ATXN7L2 in Human samples. It is validated for Western Blot.Specifications
| ATXN7L2 | |
| Polyclonal | |
| Rabbit | |
| ataxin 7-like 2, FLJ00381, MGC46534 | |
| ATXN7L2 | |
| IgG | |
| 77 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 127002 | |
| Synthetic peptides corresponding to ATXN7L2(ataxin 7-like 2) The peptide sequence was selected from the middle region of ATXN7L2. Peptide sequence NTPSPSFSKLPPSKASKSSKGKDGVEVEAPSRKRKLSPGPTTLKRTCILE. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title