missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATXN7L1 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99251H
This item is not returnable.
View return policy
Description
ATXN7L1 Polyclonal antibody specifically detects ATXN7L1 in Human samples. It is validated for Western Blot
Specifications
| ATXN7L1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 222255 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| ataxin 7-like 1, ataxin 7-like 4, ataxin-7-like protein 1, Ataxin-7-like protein 4, ATXN7L4, FLJ40255, FLJ58242, KIAA1218MGC10760, MGC33190 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human ATXN7L1. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction