missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATXN1L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA566506
This item is not returnable.
View return policy
Description
Immunogen sequence: AHSFNKAPSAT SPSGQLPHHS STQPLDLAPG RMPIYYQMSR LPAGYTLHET PPAGASPVLT PQESQS Highest antigen sequence identity to the following orthologs - mouse 87%, rat 84%.
Chromatin-binding factor that repress Notch signaling in the absence of Notch intracellular domain by acting as a CBF1 corepressor. Binds to the HEY promoter and might assist, along with NCOR2, RBPJ-mediated repression (PubMed:21475249). Can suppress ATXN1 cytotoxicity in spinocerebellar ataxia type 1 (SCA1). In concert with CIC and ATXN1, involved in brain development. [UniProt].
Specifications
| ATXN1L | |
| Polyclonal | |
| Unconjugated | |
| ATXN1L | |
| ankyrin repeat and SOCS box containing 16; ankyrin repeat and SOCS box protein 16; ankyrin repeat and SOCS box-containing 16; ankyrin repeat and SOCS box-containing protein 16; ASB16; ASB-16; ataxin 1 like; ataxin 1-like; ataxin-1-like; ATXN1L; BOAT; BOAT1; Brother of ataxin-1; brother of ATXN1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 342371 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P0C7T5 | |
| ATXN1L | |
| Recombinant Human ATXN1L. Recombinant protein control fragment (Product #RP-107176). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction