missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atuveciclib Racemate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Description
Atuveciclib Racemate (BAY-1143572 Racemate) is the racemate mixture of Atuveciclib. Atuveciclib is a potent and highly selective, oral P-TEFb/CDK9 inhibitor which supresses CDK9/CycT1 with an IC50 of 13 nM.
Specifications
Specifications
| Chemical Name or Material | Atuveciclib Racemate |
| CAS | 1414943-88-6 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C18H18FN5O2S |
| Quantity | 5 mg |
| Synonym | BAY-1143572 Racemate |
| Solubility Information | DMSO : 100 mg/mL (258.11 mM; Need ultrasonic) |
| SMILES | N=S(CC1=CC(NC2=NC(C3=CC=C(F)C=C3OC)=NC=N2)=CC=C1)(C)=O |
| Molecular Weight (g/mol) | 387.43 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction