Learn More
Invitrogen™ Attune™ Xenith™ Wash Pump Syringe
Product Code. 31759211
Change view
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. Quantity Unit Size
31759211 1 Ea. Each
1 options
Product Code. 31759211 Supplier Invitrogen™ Supplier No. 100118771CS
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
The Attune Xenith Wash Pump Syringe is part of an auxiliary pump that moves fluids through Attune Xenith Flow Cytometer for rinsing and cleaning of the system.
The Attune Xenith Wash Pump Syringe is part of an auxiliary pump that moves fluids through the Attune Xenith Flow Cytometer for rinsing and cleaning of the system. To keep your instrument in proper working order, it is important to perform regular maintenance, including changing this syringe according to the maintenance schedule laid out in the Attune Xenith user guide.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.
For Use With (Equipment) Attune Xenith Flow Cytometer
Product Type Wash Pump Syringe