Learn More
Invitrogen™ Attune™ Xenith™ Fluid Cart Spares
Product Code. 31759219
Change view
Click to view available options
Product Type:
Cart Wheel Screw
Ground Strap Screw
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
31759219 Ground Strap Screw Each
31759145 Cart Wheel Screw Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 31759219 Supplier Invitrogen™ Supplier No. 100124479CS
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
These parts are replacements of components of the Attune Xenith Flow Cytometer fluid cart.
The Invitrogen Attune Xenith Flow Cytometer is a spectrally enabled system that provides clog-resistant performance with acoustic-assisted hydrodynamic focusing. The system has an external fluid cart that holds the fluids required for everyday use of the system. These parts are replacements of various cart components.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Ea.
For Use With (Equipment) Attune Xenith Flow Cytometer
Product Type Ground Strap Screw