missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ Xenith™ Flow Cytometer

Product Code. 31662578
Change view
Click to view available options
Format:
5 mL, 2 mL and 1.5 mL Tubes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format
31662578 5 mL, 2 mL and 1.5 mL Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31662578 Supplier Invitrogen™ Supplier No. A59358
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for the rigorous demands of researchers the Attune Xenith Flow Cytometer leverages our legacy acoustic focusing technology with advanced spectral capabilities and expanded optical capabilities so users can evaluate more parameters faster.

The Attune Xenith Flow Cytometer is a reliable flow-cytometry solution designed to help meet the rigorous demands of researchers. Leveraging our legacy Attune core acoustic focusing technology, this innovative instrument combines acoustic-assisted hydrodynamic focusing with advanced spectral capabilities to help you evaluate more parameters faster. With expanded optical capabilities, including UV and NIR lasers, this system supports both traditional compensation and spectral unmixing analysis.

Features of the Attune Xenith Flow Cytometer include:

  • Remarkable speed—operates up to 5X faster than most flow cytometers
  • Versatility and flexibility—supports both spectral unmixing and conventional compensation workflows, enabling optimization of experimental design and exploration of new research possibilities
  • Compatibility with complex samples—designed to handle challenging sample types, such as tumor and tissue samples, with reduced clogging for smooth sample acquisition and reliable data analysis
  • Productivity and reliability—designed with increased uptime and simplified operation in mind, enabled by streamlined maintenance, intuitive touchscreen operation, and advanced remote diagnostics
  • Extensive detection range—includes 6 lasers, 51 fluorescent detectors, and 6 scatter channels.

Reduced time to results

Acoustic-assisted hydrodynamic focusing technology leverages ultrasonic waves to precisely align cells, delivering exceptional speed and flexibility in flow cytometry. This enables faster flow rates, while enabling uniform cell alignment in the flow cell, that help reduce the time to results for experiments and the flexibility to dilute concentrated samples without the need to spend longer times on data acquisition.

Expanded optical detection

The system includes 6 spatially separated, flat-top lasers (349, 405, 488, 561, 637, and 781 nm) and 51 fluorescent detectors with user-changeable filters as standard. It also includes 6 scatter detectors for enhanced resolution (488 nm standard FSC and SSC, 405 nm FSC and SSC for small-particle resolution, and 488 nm FSC and SSC for expanded range/polarized detection). The system can easily switch between spectral unmixing and conventional compensation workflows, making it exceptional for high-parameter analysis.

Designed to handle difficult samples

Positive displacement fluidics help reduce risk of system downtime due to clogs from sticky tumor and tissue digests. Specified volume delivery rate applies over pressure to the system that can displace clogs from the system should they occur and allow sample runs to continue.

Maintain uptime through simplified maintenance and exceptional service and support

The Attune Xenith Flow Cytometer has advanced diagnostic tools, including onboard service cameras, for assessment of flow cell cleanliness and laser alignment and onboard diagnostics and telemetry for enhanced system service logging. The integrated touchscreen can be used for easy running and monitoring of automated maintenance functions. The system includes a standard one-year warranty period. Also available are additional service plans to help take the guess work out of ensuring instrument uptime. Basic training is also provided with a member of our experienced flow cytometry support team to cover use and maintenance of the system.

TRUSTED_SUSTAINABILITY

Specifications

Format 5 mL, 2 mL and 1.5 mL Tubes
Laser Features Excitation lasers: 349nm, 405 nm, 488 nm, 561 nm, 637 nm, 781 nm
Frequency 50/60 Hz
Product Line Attune
For Use With (Equipment) Attune Xenith Flow Cytometer
Shipping Condition Room Temperature
Voltage 100/240 V
Includes Attune Xenith Flow Cytometer, Attune Xenith Fluid Cart, Minitower Windows 11 computer with instrument software, 32 inch monitor, Power cords with adapters, Fiber Ethernet cable, Attune Shutdown solution, Attune Wash solution, Attune Focusing Fluid, Attune NxT Flow cell cleaning solution, Attune Debubble solution, Attune Xenith Quality Control Beads

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.