missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Attune™ NxT Waste Bottle
Click to view available options
Quantity:
1 Each
Unit Size:
Each
Description
This is a 1.9 L replacement bottle for the waste tank on the Attune™ NxT Acoustic Focusing Cytometer.
Specifications
Specifications
| Content And Storage | Room temperature |
| Quantity | 1 Each |
| For Use With (Equipment) | Attune Acoustic Focusing Cytometer |
| Product Type | Attune™ NxT Waste Bottle |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction