missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT Small Particle Side-Scatter Filter

Product Code. 16364972
Click to view available options
Quantity:
1 filter
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16364972

Brand: Invitrogen™ 100083194

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Attune NxT Small Particle Side-Scatter Filter is for use with the Attune NxT Acoustic Focusing Cytometer.

It is designed to increase the dynamic range of the side-scatter detection without sacrificing resolution of the detector. The use of this 488/10 side scatter filter combined with ultra-filtering of the focusing fluid has been found to dramatically decrease background noise and enable discrimination of 100-nm particles.

Easy to use
This filter is easily swapped for the standard side-scatter filter. It cannot be installed the wrong way because the optical block is keyed to accept only one orientation of the filter blade. The filter also has a color-coded label to help identify the correct position.

TRUSTED_SUSTAINABILITY

Specifications

Product Line Attune
Quantity 1 filter
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Block Format Non-Interchangeable
For Use With (Equipment) Attune Acoustic Focusing Cytometer
Product Type Small Particle Side-Scatter Filter

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.