missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Attune™ NxT Focus BTL Tubing

Product Code. 17166088
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17166088 1 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17166088 Supplier Invitrogen™ Supplier No. 100096711CST
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

This is replacement tubing for the focusing fluid bottle of the Attune NxT Acoustic Focusing Cytometer

  • This is replacement tubing for the focusing fluid bottle of the Attune NxT Acoustic Focusing Cytometer.
  • The tubing can be replaced easily without replacing the entire bottle assembly if the tubing connector to the instrument is broken.
TRUSTED_SUSTAINABILITY

Specifications

Product Line FPL
Includes 1 tubing set
Quantity 1 each
Format Tubing
Shipping Condition Room temperature
For Use With (Equipment) Attune NxT
Product Type Tubing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.