Learn More
Invitrogen™ Attune™ NxT Custom Filter Holder Kit
Allows users of the Attune™ NxT acoustic focusing cytometer to make their own custom emission or dichroic filters in order to customize the optical configuration of the instrument
Brand:  Invitrogen™ A27784
Additional Details : Weight : 1.00000kg
Product Code. 15833374
  • £57.75 / 1 set
Estimated Shipment: 12-08-2024
to see stock.
Description
Description
  • The Attune™ NxT Custom Filter Holder Kit allows users of the Attune™ NxT acoustic focusing cytometer to make their own custom emission or dichroic filters in order to customize the optical configuration of the instrument.
  • Each kit contains sufficient parts to make 2 emission filters and 2 dichroic filters.
  • The kit includes instructions for attaching the filter to the filter blade, but does not include the glass filter.
Specifications
Specifications
Attune NxT Acoustic Focusing Cytometer
Custom Filter Holder Kit
1 Kit
2 dichroic filter blades
2 emission filter blades
4 clips
Instruction card
Approved for shipment at Room Temperature or on Wet Ice
Attune™