missing translation for 'onlineSavingsMsg'
Learn More

Attune™ NxT Automation SmartStor Sample Management System

Product Code. 32096683
Change view
Click to view available options
Quantity:
1 system
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
32096683 1 system Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32096683 Supplier Invitrogen Supplier No. A35221
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket

Specifications

Product Line Attune
Quantity 1 system
Shipping Condition Room Temperature
Block Format Non-Interchangeable
For Use With (Equipment) Attune NxT Acoustic Focusing Cytometer
Product Type Sample Management System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.