missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Attune™ CytPix™ Violet 6 Conversion Kit, for blue laser
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
Description
- Designed to reconfigure Attune NxT four-channel violet instruments to allow detection of six channels of violet
- A conversion kit is required for each original, and for any upgraded lasers of the instrument
- Conversion is carried out by a field service engineer
Specifications
Specifications
| For Use With (Application) | Flow Cytometry |
| For Use With (Equipment) | Attune CytPix Flow Cytometer |
| Includes | Filters, dichroics |
| Product Line | DJM |
| Quantity | 1 Kit |
| Regulatory Status | RUO |
| Shipping Condition | RT |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction